Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL25A1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89114858 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-114-858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-114-858 Supplier Abnova Corporation Supplier No. PAB1057

Rabbit polyclonal antibody raised against synthetic peptide of COL25A1.

COL25A1 is a brain-specific membrane-bound collagen. Proteolytic processing releases CLAC, a soluble form of COL25A1 containing the extracellular collagen domains that associates with senile plaques in Alzheimer disease (AD; MIM 104300) brains (Osada et al., 2005 [PubMed 15615705]).[supplied by OMIM

Sequence: DYNGNLHEALQRIT

Specifications

Antigen COL25A1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description collagen, type XXV, alpha 1
Formulation In PBS (0.02% sodium azide)
Gene COL25A1
Gene Alias CLAC, CLACP
Gene Symbols COL25A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 430-443 of human COL25A1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84570
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.