Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COL2A1 Rabbit anti-Chicken, Polyclonal Antibody, Abnova™

Catalog No. 89116704 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-704 Supplier Abnova Corporation Supplier No. PAB13495
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native COL2A1.

Specifications

Antigen COL2A1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native COL2A1.
Dilution Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from phosphate buffer.
Gene COL2A1
Gene Symbols COL2A1
Host Species Rabbit
Immunogen Native purified chicken cartilage COL2A1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 395069
Test Specificity This antibody reacts with chicken collagen type II 100%, no crossreactivity with chicken collagen type I and III, no crossreactivity with human, mouse and rat collagen type II.
Target Species Chicken
Content And Storage Store at -20°C.
Avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.