Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-COL2A1, Polyclonal, Abnova™

Catalog No. 89120482 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-120-482 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-120-482 Supplier Abnova Corporation Supplier No. PAB18923

Specifications

Antigen COL2A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene Symbols COL2A1
Host Species Rabbit
Immunogen Native purified COL2A1 from human knee cartilage and bovine tissues.
Quantity 1 mL
Primary or Secondary Primary
Gene ID (Entrez) 1280
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.