Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen I and III Rabbit anti-Dog, Polyclonal Antibody, Abnova™

Catalog No. 89116817 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-817 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-817 Supplier Abnova Corporation Supplier No. PAB13715
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Collagen I and III.

Specifications

Antigen Collagen I and III
Applications Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Collagen I and III.
Dilution Immunohistochemistry (Frozen sections) (1:10-1:20) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from purified antibody
Gene Symbols COL1A1
Host Species Rabbit
Immunogen Native purified dog Collagen I and III.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403651/403824/478835
Test Specificity Recognizes dog collagen type I and III.
Target Species Dog
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.