Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type IV Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89137608 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-608 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-608 Supplier Abnova Corporation Supplier No. PAB13502
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Collagen Type IV.

Specifications

Antigen Collagen Type IV
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Collagen Type IV.
Dilution Immunohistochemistry (1:500-1:1000) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from phosphate buffer
Host Species Rabbit
Immunogen Native purified Collagen Type IV from human placenta.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibory reacts with human collagen type IV 100%; human collagen I, II <0.1%; human collagen III, V <1.0%; human fibronectin <0.1%; mouse laminin <0.1% (RIA at 1:10000 dilution).
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.