Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VI Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89137612 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-612 Supplier Abnova Corporation Supplier No. PAB13510
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Collagen Type VI.

Specifications

Antigen Collagen Type VI
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Collagen Type VI.
Dilution ELISA (1:200) Immunohistochemistry (1:400-1:1000) Immunofluorescence (1:80) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from phosphate buffer
Host Species Rabbit
Immunogen Native purified Collagen Type VI from human placenta.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with human collagen type VI 100% binding in RIA at 1:10.000 dilution, cross-reactivity with human collagen type I, II, III, IV and V <0.1%; human elastin and human laminin <0.1%; human fibronectin <1%.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.