Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Collagen Type VI Rabbit anti-Bovine, Human, Polyclonal Antibody, Abnova™

Catalog No. 89136943 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-943 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-943 Supplier Abnova Corporation Supplier No. PAB10195
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against native Collagen Type VI.

Specifications

Antigen Collagen Type VI
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Collagen Type VI.
Dilution Sandwich ELISA (1:4000-1:8000) Western Blot (1:5000-1:10000) Immunohistochemistry (Frozen sections) (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In 100mM Na2B4O7, 75mM NaCl, 5mM EDTA, pH 8.0 (0.01% sodium azide)
Host Species Rabbit
Immunogen Native purified human and bovine placenta Collagen Type VI.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with most mammalian Type VI collagens and has negligible cross-reactivity with Type I, II, III, IV or V collagens. Non-specific cross-reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible.
Target Species Bovine, Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.