Learn More
Description
Specifically detects COPA in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
| Antigen | COPA |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Formulation | Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.02% sodium azide and 40% glycerol. |
| Gene Accession No. | P53621 |
| Gene Symbols | COPA |
| Host Species | Rabbit |
| Immunogen | Recombinant protein corresponding to human COPA. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.