Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COPZ1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COPZ1.
Supplier: Abnova Corporation PAB26995
Specifications
COPZ1 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
COPZ1 | |
CGI-120/COPZ/zeta1-COP | |
Rabbit | |
Antigen affinity purification | |
RUO | |
22818 | |
Human, Mouse | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of COPZ1. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
P61923 | |
COPZ1 | |
A synthetic peptide corresponding to human COPZ1. | |
100 μL | |
Primary | |
COPZ1 polyclonal antibody detects endogenous levels of COPZ1 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction