Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corin Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Corin.
Supplier: Abnova Corporation PAB12762
Specifications
Corin | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Corin. | |
In TBS, pH 7.2 (BSA, 10% Proclin300) | |
AV273130/Lrp4/MGC118735 | |
Rabbit | |
100 μg | |
Primary | |
This antibody recognizes ∼120 KDa of mouse Corin. | |
Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
Corin | |
Corin | |
A synthetic peptide corresponding to N-terminus of mouse Corin. | |
RUO | |
53419 | |
Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction