Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-COUP-TF2, Polyclonal,

Catalog No. ABE1426MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE1426MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1426MI Supplier MilliporeSigma™ Supplier No. ABE1426

Rabbit Polyclonal Antibody

Specifically detects COUP-TF2 in Human, Mouse samples, and it is validated for ChIP, Immunofluorescence, Immunohistochemistry, Western Blotting

COUP transcription factor 2 (UniProt P43135; also known as Apolipoprotein AI regulatory protein 1, Apolipoprotein regulatory protein 1, ARP-1, COUP-TF II, COUP transcription factor II, Nuclear receptor subfamily 2 group F member 2) is encoded by the Nr2f2 (also known as Aporp1, Arp-1, Arp1, Tfcoup2) gene (Gene ID 11819) in murine species. COUP transcription factors1&2 (COUP-TFI & COUP-TFII) are two related nuclear orphan receptors implicated in their roles in guiding migrating neurons towards their target regions during brain development. COUP-TFI and COUP-TFII are expressed in distinct and non-overlapping migrating cells in specific pathways in the basal telencephalon of mouse embryos at a stage when interneuron migration is robust. While COUP-TFI appears to control dorsal and ventral migration in the basal telencephalon, COUP-TFII is more specific in modulating tangential migration into the intermediate zone of the cortex. Coup-TFII is also identified as an activator of Prox1 expression in venous lymphatic endothelial cell (LEC) progenitors during early stages of LEC specification and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COUP-TF2
Applications ChIP Assay, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. P43135
Gene Symbols Nr2f2; Aporp1; Arp-1; Arp1; Tfcoup2
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the the N-terminal half of mouse COUP-TF2.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_033827
Target Species Human, Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.