Learn More
COX6C, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of COX6C.
Supplier: Abnova Corporation PAB17522
Description
Cytochrome c oxidase (COX), the terminal enzyme of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. It is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may be involved in the regulation and assembly of the complex. This nuclear gene encodes subunit VIc, which has 77% amino acid sequence identity with mouse COX subunit VIc. This gene is up-regulated in prostate cancer cells. A pseudogene COX6CP1 has been found on chromosomes 16p12. [provided by RefSeq
Specifications
COX6C | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of COX6C. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
P09669 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total COX6C protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
COX6C | |
COX6C | |
A synthetic peptide corresponding to internal of human COX6C. | |
RUO | |
1345 | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only