Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CR1, Rabbit, Clone: SP197, Abnova™

Catalog No. 89110308 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-308 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-308 Supplier Abnova Corporation Supplier No. MAB12167

Rabbit monoclonal antibody raised against synthetic peptide of human CR1.

This gene is a member of the receptors of complement activation (RCA) family and is located in the 'cluster RCA' region of chromosome 1. The gene encodes a monomeric single-pass type I membrane glycoprotein found on erythrocytes, leukocytes, glomerular podocytes, and splenic follicular dendritic cells. The Knops blood group system is a system of antigens located on this protein. The protein mediates cellular binding to particles and immune complexes that have activated complement. Decreases in expression of this protein and/or mutations in its gene have been associated with gallbladder carcinomas, mesangiocapillary glomerulonephritis, systemic lupus erythematosus and sarcoidosis. Mutations in this gene have also been associated with a reduction in Plasmodium falciparum rosetting, conferring protection against severe malaria. Alternate allele-specific splice variants, encoding different isoforms, have been characterized. Additional allele specific isoforms, including a secreted form, have been described but have not been fully characterized. [provided by RefSeq]

Specifications

Antigen CR1
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP197
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human CR1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (Tissue lysate) (1:400) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene CR1
Gene Alias C3BR/CD35/KN
Gene Symbols CR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CR1.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1378
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.