Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CR2, Rabbit, Clone: SP186, Abnova™

Catalog No. 89110302 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-302 Supplier Abnova Corporation Supplier No. MAB12161

Rabbit monoclonal antibody raised against synthetic peptide of human CR2.

Complement component receptor-2 (CR2) is the membrane protein on B lymphocytes to which the Epstein-Barr virus (EBV) binds during infection of these cells. See also CR1 (MIM 120620). Yefenof et al. (1976) [PubMed 181330] found complete overlapping of EBV receptors and C3 (MIM 120700) receptors on human B lymphocytes.[supplied by OMIM

Specifications

Antigen CR2
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone SP186
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of human CR2.
Dilution Flow Cytometry (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (Cell lysate) (1:400) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.6 (1% BSA, 0.09% sodium azide)
Gene CR2
Gene Alias C3DR/CD21/SLEB9
Gene Symbols CR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus region of human CR2.
Purification Method Protein A/G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1380
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.