Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRB2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128864 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-864 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-864 Supplier Abnova Corporation Supplier No. PAB27210
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CRB2.

Specifications

Antigen CRB2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CRB2.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CRB2
Gene Accession No. NP_775960
Gene Alias FLJ16786/FLJ38464
Gene Symbols CRB2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 17 amino acids at internal region of human CRB2.
Purification Method Peptide affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286204
Test Specificity At least three isoforms of CRB2 are known to exist; this antibody will detect all three. CRB2 antibody is predicted to not cross-react with other members of the CRB family.
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.