Learn More
CREB1 (phospho S142), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CREB1.
Supplier: Abnova Corporation PAB25875
Description
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: D-L-Sp-S-DSpecifications
CREB1 | |
Polyclonal | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
CREB1 | |
CREB/MGC9284 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1385 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CREB1. | |
In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
P16220 | |
CREB1 | |
Synthetic phosphopeptide corresponding to residues surrounding S142 of human CREB1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only