Learn More
CREB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Specifications
Specifications
| Antigen | CREB1 |
| Applications | ELISA, Gel Shift, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against synthetic peptide of CREB1. |
| Dilution | Western Blot (1:500-1:1000) Gel Supershift Assay (0.5-1.0 ul per assay) The optimal working dilution should be determined by the end user. |
| Formulation | In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) |
| Gene | CREB1 |
| Gene Alias | CREB/MGC9284 |
| Gene Symbols | CREB1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.