Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB3L4, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121349 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-349 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-349 Supplier Abnova Corporation Supplier No. PAB19819
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CREB3L4.

cAMP response element-binding (CREB) proteins are a family of mammalian transcription activators. For further background information on CREB proteins, see CREB1 (MIM 123810).[supplied by OMIM

Sequence: PSGRIRSVLHADEM

Specifications

Antigen CREB3L4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CREB3L4.
Dilution ELISA (1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user.
Formulation In serum (0.02% sodium azide)
Gene CREB3L4
Gene Accession No. Q8TEY5
Gene Alias AIBZIP/ATCE1/CREB3/CREB4/JAL/hJAL
Gene Symbols CREB3L4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CREB3L4.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148327
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.