Learn More
CRHR1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CRHR1.
Supplier: Abnova Corporation PAB26102
Description
This gene encodes a G-protein coupled receptor that binds neuropeptides of the corticotropin releasing hormone family that are major regulators of the hypothalamic-pituitary-adrenal pathway. The encoded protein is essential for the activation of signal transduction pathways that regulate diverse physiological processes including stress, reproduction, immune response and obesity. Alternative splicing results in multiple transcript variants
Specifications
CRHR1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CRHR1. | |
In PBS (0.09% sodium azide) | |
P34998 | |
CRHR1 | |
A synthetic peptide corresponding to 17 amino acids at N-terminus of human CRHR1. | |
50 μg | |
Primary | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (13 ug/mL) The optimal working dilution should be determined by the end user. | |
CRHR1 | |
CRF-R/CRF1/CRFR1/CRH-R1h/CRHR/CRHR1f | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
1394 | |
Human, Rat | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only