Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cricket paralysis virus CrPV Genome polyprotein, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cricket paralysis virus CrPV Genome polyprotein.
Supplier: Abnova Corporation PAB15962
Specifications
Cricket paralysis virus CrPV Genome polyprotein | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Cricket paralysis virus CrPV Genome polyprotein. | |
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
A synthetic peptide corresponding to C-terminus of Cricket paralysis virus CrPV Genome polyprotein. | |
RUO | |
Virus | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
Rabbit | |
100 μg | |
Primary | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction