Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIM1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89127117 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-117 Supplier Abnova Corporation Supplier No. PAB25500
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CRIM1.

Motor neurons are among the earliest neurons to appear after the commencement of cell patterning and the beginning of cell differentiation. Differentiation occurs in a ventral-to-dorsal gradient and is mediated, at least in part, by the concentration of ventrally expressed Sonic hedgehog protein (SHH; MIM 600725). Dorsally expressed factors, such as members of the bone morphogenic protein (e.g., BMP4; MIM 112262) and transforming growth factor-beta (e.g., TGFB1; MIM 190180) families, can repress the induction of these neurons. CRIM1 may interact with growth factors implicated in motor neuron differentiation and survival (Kolle et al., 2000 [PubMed 10642437]).[supplied by OMIM

Specifications

Antigen CRIM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CRIM1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CRIM1
Gene Accession No. NP_057525
Gene Alias MGC138194/S52
Gene Symbols CRIM1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CRIM1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51232
Test Specificity CRIM1 antibody is human reactive. CRIM1 antibody is predicted to not cross-react with CRIM2.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.