Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRNN, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116841 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-841 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-841 Supplier Abnova Corporation Supplier No. PAB13784
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CRNN.

This gene encodes a member of the fused gene family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation. [provided by RefSeq]

Sequence: EATNDQNRGTETHGQG

Specifications

Antigen CRNN
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CRNN.
Dilution Western Blot (1:2000) Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.01% sodium azide)
Gene CRNN
Gene Alias C1orf10/DRC1/PDRC1/SEP53
Gene Symbols CRNN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 259-274 of human CRNN.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49860
Test Specificity Recognizes human cornulin. Detects a band of ∼70 KDa by Western blot.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.