Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2RB Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89131415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-131-415 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-131-415 Supplier Abnova Corporation Supplier No. PAB4461
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CSF2RB.

The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP). [provided by RefSeq

Specifications

Antigen CSF2RB
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CSF2RB.
Dilution ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CSF2RB
Gene Accession No. NP_000386;P32927
Gene Alias CD131/CDw131/IL3RB/IL5RB
Gene Symbols CSF2RB
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus (V759) of human CSF2RB.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1439
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.