Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTGF, Rabbit, Biotin, Polyclonal Antibody, Abnova™

Catalog No. 89117411 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-411 Supplier Abnova Corporation Supplier No. PAB15040
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CTGF.

Specifications

Antigen CTGF
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Description Rabbit polyclonal antibody raised against synthetic peptide of CTGF.
Dilution Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene CTGF
Gene Alias CCN2/HCS24/IGFBP8/MGC102839/NOV2
Gene Symbols CTGF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 299-349 of human CTGF.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1490
Test Specificity This antibody reacts with murine CTGF protein. The immunogen used for this antibody production has 100% homology with human, rat, and mouse protein. This is specific to CTGF. By Western blot a band at ∼38 KDa is seen. No other applications have been tested.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.