Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rabbit anti-CTHRC1, Clone: Vli-55,
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma™ MABT889
Description
Specifically detects CTHRC1 clone: Vli-55 in Human, Mouse, Porcine, Rat samples, and it is validated for Immunohistochemistry, Western Blotting
Collagen triple helix repeat-containing protein 1 (UniProt Q96CG8; also known as Protein NMTC1) is encoded by the CTHRC1 gene (ORF UNQ762/PRO1550; Gene ID 115908) in human. CTHRC1 is a secreted glycoprotein with 12 repeats of the Gly-X-Y motif (a.a. 58-93 based on the pro-form seqeunce) that is reported to increase cell motility and promote cell migration by limiting the deposition of collagen matrix. CTHRC1 is normally not expressed in blood vessels, but is induced in adventitial cells in remodeling arteries, as well as in dermal fibroblasts during wound healing. Upregulatetd CTHRC1 level is also observed in many human solid tumors, including non-small-cell lung cancer (NSCLC), gastric cancer, hepatocellular cancer, breast cancer, pancreatic cancer, and colorectal cancer (CRC). CTHRC1 is initially produced with a signal peptide sequence (a.a. 1-30), the removal of which yields the 213-amino acid (a.a. 31-243) mature protein.
Specifications
CTHRC1 | |
Monoclonal | |
Purified rabbit monoclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
CTHRC1; UNQ762; PRO1550 | |
Linear peptide corresponding to a C-terminal sequence of human CTHRC1. | |
200 μg | |
Cell Structure | |
NP_612464.1 | |
Stable for one year at 2°C to 8°C from date of receipt. |
Immunohistochemistry, Western Blot | |
Vli-55 | |
Q96CG8 | |
Rabbit | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse, Pig, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction