Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CTNND1, Rabbit, Clone: SP63, Abnova™

Catalog No. 89108338 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-338 Supplier Abnova Corporation Supplier No. MAB10024

Rabbit monoclonal antibody raised against synthetic peptide of CTNND1.

This gene encodes a member of the Armadillo protein family, which function in adhesion between cells and signal transduction. Multiple translation initiation codons and althernative splicing result in many different isoforms being translated. Not all of the full-length natures of the described transcript variants have been determined. [provided by RefSeq

Specifications

Antigen CTNND1
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone SP63
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against synthetic peptide of CTNND1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (BSA, 0.09% sodium azide)
Gene CTNND1
Gene Alias CAS/CTNND/KIAA0384/P120CAS/P120CTN/p120
Gene Symbols CTNND1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CTNND1.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1500
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.