Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctr9, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89116916 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-916 Supplier Abnova Corporation Supplier No. PAB14078
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Ctr9.

Specifications

Antigen Ctr9
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Ctr9.
Dilution ELISA (1:100-1:500) Western Blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In serum (0.05% sodium azide)
Gene Ctr9
Gene Accession No. Q6PD62 (human)
Gene Alias AA409336/KIAA0155/Sh2bp1/Tsbp/Tsp/mKIAA0155
Gene Symbols Ctr9
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to N-terminus of mouse Ctr9.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22083
Target Species Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.