Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CUL1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89128693 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-128-693 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-128-693 Supplier Abnova Corporation Supplier No. PAB27044
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CUL1.

Specifications

Antigen CUL1
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CUL1.
Dilution Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene CUL1
Gene Accession No. Q13616
Gene Alias MGC149834/MGC149835
Gene Symbols CUL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CUL1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8454
Test Specificity CUL1 polyclonal antibody detects endogenous levels of CUL1 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.