Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CUL5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CUL5.
Supplier: Abnova Corporation PAB10039
Specifications
CUL5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of CUL5. | |
In antiserum (0.01% sodium azide) | |
AAR13072 | |
CUL5 | |
A synthetic peptide (conjugated with KLH) corresponding to amino acids 4-18 of human CUL5. | |
RUO | |
8065 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
CUL5 | |
VACM-1/VACM1 | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction