Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CX3CL1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89116119 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-116-119 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-116-119 Supplier Abnova Corporation Supplier No. PAB12834
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CX3CL1.

Specifications

Antigen CX3CL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CX3CL1.
Dilution Western Blot (0.5-1 mg/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene CX3CL1
Gene Accession No. AAB50014
Gene Alias ABCD-3/C3Xkine/CXC3/CXC3C/NTN/NTT/SCYD1/fractalkine/neurotactin
Gene Symbols CX3CL1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to N-terminus of human CX3CL1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6376
Target Species Human, Mouse
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.