Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CXCR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89134036 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-036 Supplier Abnova Corporation Supplier No. PAB8941
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CXCR1.

The protein encoded by this gene is a member of the G-protein-coupled receptor family. This protein is a receptor for interleukin 8 (IL8). It binds to IL8 with high affinity, and transduces the signal through a G-protein activated second messenger system. Knockout studies in mice suggested that this protein inhibits embryonic oligodendrocyte precursor migration in developing spinal cord. This gene, IL8RB, a gene encoding another high affinity IL8 receptor, as well as IL8RBP, a pseudogene of IL8RB, form a gene cluster in a region mapped to chromosome 2q33-q36. [provided by RefSeq

Sequence: DDLNFTGNPPADEDYSPC

Specifications

Antigen CXCR1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CXCR1.
Dilution Western Blot (1:500) Immunoprecipitation (1:250) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.02% sodium azide
Gene CXCR1
Gene Alias C-C/C-C-CKR-1/CD128/CD181/CDw128a/CKR-1/CMKAR1/IL8R1/IL8RA/IL8RBA
Gene Symbols CXCR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 13-30 of human CXCR1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3577
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.