Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYP19A1 Rabbit anti-Bovine, Horse, Human, Primates, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115827 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-827 Supplier Abnova Corporation Supplier No. PAB12453
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CYP19A1.

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and catalyzes the last steps of estrogen biosynthesis, three successive hydroxylations of the A ring of androgens. Mutations in this gene can result in either increased or decreased aromatase activity; the associated phenotypes suggest that estrogen functions both as a sex steroid hormone and in growth or differentiation. The gene expresses two transcript variants. [provided by RefSeq

Specifications

Antigen CYP19A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CYP19A1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene CYP19A1
Gene Accession No. P11511
Gene Alias ARO/ARO1/CPV1/CYAR/CYP19/MGC104309/P-450AROM
Gene Symbols CYP19A1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 400-502 of human CYP19A1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1588
Test Specificity This antibody is specific to aromatase.
Target Species Bovine, Equine, Human, Primate, Rabbit
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.