Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYSLTR1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™

Catalog No. 89127814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-127-814 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-127-814 Supplier Abnova Corporation Supplier No. PAB26174
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CYSLTR1.

The cysteinyl leukotrienes LTC4, LTD4, and LTE4 are important mediators of human bronchial asthma. Pharmacologic studies have determined that cysteinyl leukotrienes activate at least 2 receptors, the protein encoded by this gene and CYSLTR2. This encoded receptor is a member of the superfamily of G protein-coupled receptors. Activation of this receptor by LTD4 results in contraction and proliferation of smooth muscle, oedema, eosinophil migration and damage to the mucus layer in the lung. [provided by RefSeq

Specifications

Antigen CYSLTR1
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CYSLTR1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (6-14 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CYSLTR1
Gene Accession No. Q9Y271
Gene Alias CYSLT1/CYSLT1R/CYSLTR/HG55/HMTMF81/MGC46139
Gene Symbols CYSLTR1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to 20 amino acid at extracellular domain of human CYSLTR1.
Purification Method Immunoaffinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10800
Test Specificity BLAST analysis of the peptide immunogen showed no homology with other human proteins, except NMU (45%).
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.