Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytip, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89121449 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-121-449 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-121-449 Supplier Abnova Corporation Supplier No. PAB19941
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Cytip.

Specifications

Antigen Cytip
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cytip.
Dilution ELISA (1:5000-1:20000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 0.02M KH2PO4, 0.15M NaCl, pH 7.2 (0.01% sodium azide)
Gene Cytip
Gene Alias A130053M09Rik/AI462064/C80816/Cbp/Pscdbp
Gene Symbols Cytip
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of mouse Cytip.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227929
Test Specificity Expect a band approximately 40 kDa in size corresponding to Cybr by western blotting in the appropriate cell lysate or extract.
Target Species Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.