Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytomegalovirus (AD 169), Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89137744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-744 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-744 Supplier Abnova Corporation Supplier No. PAB13933

Rabbit polyclonal antibody raised against Cytomegalovirus (AD 169).

Specifications

Antigen Cytomegalovirus (AD 169)
Applications ELISA, Immunoblot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against Cytomegalovirus (AD 169).
Dilution The optimal working dilution should be determined by the end user.
Formulation In antiserum
Host Species Rabbit
Immunogen Cytomegalovirus strain AD169, whole virus.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Reacts with various antigens from all CMV strains. No cross reactivity with other herpes viruses.
Target Species Cytomegalovirus (CMV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.