Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CYTSB Rabbit anti-Human, DyLight 649, Polyclonal Antibody, Abnova™

Catalog No. 89117386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-386 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-386 Supplier Abnova Corporation Supplier No. PAB14980
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of CYTSB.

Specifications

Antigen CYTSB
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 649
Description Rabbit polyclonal antibody raised against synthetic peptide of CYTSB.
Dilution Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In 50mM sodium borate
Gene CYTSB
Gene Alias FLJ36955/HCMOGT-1/NSP/SPECC1
Gene Symbols CYTSB
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of human CYTSB.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92521
Test Specificity This antibody reacts with human protein. There is 93% homology of the immunogen with the mouse protein and 87% homology of the immunogen with the rat NSP5a3a protein. This is specific to NSP5a3a. This antibody is useful for Western blot analysis, where a band is seen ∼80 KDa. A second band may be seen ∼60 KDa. The specificity of this band is not known. No other applications have been tested. Optimal working dilutions should be determined by the investigator.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.