Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
D-Serine, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against D-Serine.
Supplier: Abnova Corporation PAB14719
Specifications
D-Serine | |
Polyclonal | |
Rabbit polyclonal antibody raised against D-Serine. | |
Lyophilized from PBS, pH 7.4 | |
D-Serine conjugated with carrier protein. | |
100 μL | |
Primary | |
Lyophilized |
ELISA, Immunohistochemistry, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
ELISA (1:2000-1:10000) Immunohistochemistry (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
Store at 4°C on dry atmosphere. After reconstitution with 50μL of distilled water and 50μL of glycerol store at -20°C for 2 years. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction