Learn More
DAPK3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DAPK3.
Supplier: Abnova Corporation PAB18339
Description
Death-associated protein kinase 3 (DAPK3) induces morphological changes in apoptosis when overexpressed in mammalian cells. These results suggest that DAPK3 may play a role in the induction of apoptosis. [provided by RefSeq
Specifications
DAPK3 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user. | |
DAPK3 | |
FLJ36473/ZIP/ZIPK | |
Rabbit | |
Affinity purification | |
RUO | |
1613 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of DAPK3. | |
In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
O43293 | |
DAPK3 | |
A synthetic peptide corresponding to residues surrounding T265 of human DAPK3. | |
100 μg | |
Primary | |
This antibody is specific to DAPK3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only