Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Decorin, KLH-conjugated, Polyclonal,

Catalog No. ABT276MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT276MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT276MI Supplier MilliporeSigma™ Supplier No. ABT276
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Decorin in Human, Monkey samples, and it is validated for Immunohistochemistry, Western Blotting

Decorin (UniProt P07585), also known as Bone proteoglycan II, PG40, PG-S2) is encoded by the DCN gene in human. Decorin is a pericellular matrix proteoglycan structurally related to two other small proteoglycans, biglycan and fibromodulin. Decorin and biglycan are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while biglycan contains two chains. Decorin modulates collagen fibril formation as well as the bioactivity of various members of the transforming growth factor-beta (TGFbeta) superfamily. Decorin is initially translated as a pre-proprotein with an N-terminal signal peptide (aa 1-16) and propeptide (aa 17-30) sequence. Sequential removal of the signal peptide and the propeptide sequence produces pro- and mature decorin, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Decorin
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Formulation Rabbit polyclonal antibody serum with 0.05% sodium azide.
Gene Accession No. P07585
Gene Symbols DCN; SLRR1B
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the mature chain of human Decorin.
Purification Method Unpurified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001911
Target Species Human, Monkey
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.