Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dengue Virus Capsid Protein Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP242823
Description
Specifically detects Dengue Virus Capsid Protein in Hamster, Virus samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting
Dengue Virus Capsid Protein Polyclonal specifically detects Dengue Virus Capsid Protein in Viral samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Dengue Virus Capsid Protein | |
Polyclonal | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:2000 | |
POLY | |
Full length Dengue virus 2 Capsid Recombinant protein (Dengue virus 2 (strain 16681 PDK 53)) | |
Affinity Purified | |
RUO | |
Primary | |
Virus | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Rabbit | |
12 kDa | |
0.1 mL | |
Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
1494449 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction