Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DES Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89115455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-455 Supplier Abnova Corporation Supplier No. PAB11919

Rabbit polyclonal antibody raised against synthetic peptide of DES.

This gene encodes a muscle-specific class III intermediate filament. Homopolymers of this protein form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. Mutations in this gene are associated with desmin-related myopathy, a familial cardiac and skeletal myopathy (CSM), and with distal myopathies. [provided by RefSeq

Specifications

Antigen DES
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration Lot specific
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DES.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Gene DES
Gene Alias CMD1I/CSM1/CSM2/FLJ12025/FLJ39719/FLJ41013/FLJ41793
Gene Symbols DES
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human DES.
Purification Method Immunoaffinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1674
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.