Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dffa Rabbit anti-Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89117091 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-117-091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-117-091 Supplier Abnova Corporation Supplier No. PAB14474
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of Dffa.

Specifications

Antigen Dffa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Dffa.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene Dffa
Gene Accession No. O54786
Gene Alias A330085O09Rik/DFF35/Dff45/ICAD/ICAD-L/ICAD-S
Gene Symbols Dffa
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-21 of mouse Dffa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13347
Test Specificity This antibody recognizes non-cleaved (45 kD) and cleaved ICAD. Eptiopes at the N-terminus of mouse ICAD.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.