Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DFNB31, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89130184 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-184 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-184 Supplier Abnova Corporation Supplier No. PAB28671

Rabbit polyclonal antibody raised against recombinant DFNB31.

In rat brain, Cip98 interacts with a calmodulin-dependent serine kinase, CASK (MIM 300172), and may be involved in the formation of scaffolding protein complexes that facilitate synaptic transmission in the central nervous system (CNS) (Yap et al., 2003 [PubMed 12641734]). Mutations in this gene, also known as WHRN, cause DFNB31 (MIM 607084).[supplied by OMIM

Sequence: VDPGSEAEGSGLKVGDQILEVNGRSFLNILHDEAVRLLKSSRHLILTVKDVGRLPHARTTVDETKWIASSRIRETMANSAGFLGDLTTEGINKPGFYKGPAGSQVTLSSLGNQTRVLLEEQARHLLNEQEH

Specifications

Antigen DFNB31
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant DFNB31.
Dilution Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Gene DFNB31
Gene Alias CIP98/DKFZp434N014/KIAA1526/RP11-9M16.1/USH2D/WHRN/WI
Gene Symbols DFNB31
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids of human DFNB31.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25861
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.