Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGAT1, Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89115823 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-115-823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-115-823 Supplier Abnova Corporation Supplier No. PAB12449
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DGAT1.

The enzyme encoded by this gene utilizes diacylglycerol and fatty acyl CoA as substrates in order to catalyze the final stage of triacylglycerol synthesis. It is also involved in cellular as well as physiological metabolic processes. [provided by RefSeq

Specifications

Antigen DGAT1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DGAT1.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene DGAT1
Gene Accession No. O75907
Gene Alias ARGP1/DGAT
Gene Symbols DGAT1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 200-300 of human DGAT1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8694
Target Species Bovine, Fish, Goat, Human, Mouse, Primate, Rat, Sheep
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.