Learn More
DGKH, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DGKH.
Supplier: Abnova Corporation PAB17407
Description
This gene encodes a member of the diacylglycerol kinase (DGK) enzyme family of proteins, specifically the type II DGK subfamily. Members of this family are involved in regulating the intracellular concentrations of diacylglycerol and phosphatidic acid. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
DGKH | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of DGKH. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
Q86XP1 | |
DGKH | |
A synthetic peptide corresponding to internal of human DGKH. | |
RUO | |
160851 | |
Human, Mouse | |
Liquid |
ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
DGKH | |
DGKeta/DKFZp761I1510 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total DGKH protein. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only