Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DGKQ Rabbit anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89130960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-130-960 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-130-960 Supplier Abnova Corporation Supplier No. PAB3339
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against synthetic peptide of DGKQ.

The protein encoded by this gene contains three cysteine-rich domains, a proline-rich region, and a pleckstrin homology domain with an overlapping Ras-associating domain. It is localized in the speckle domains of the nucleus, and mediates the regeneration of phosphatidylinositol (PI) from diacylglycerol in the PI-cycle during cell signal transduction. [provided by RefSeq

Specifications

Antigen DGKQ
Applications Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of DGKQ.
Dilution Flow Cytometry (1:10-50) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:10-50) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene DGKQ
Gene Accession No. P52824
Gene Alias DAGK/DAGK4/DAGK7
Gene Symbols DGKQ
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human DGKQ.
Purification Method Protein A purification
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1609
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.