Learn More
DHX40, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant DHX40.
Supplier: Abnova Corporation PAB24320
Description
DDX40 is a member of the DExH/D box family of ATP-dependent RNA helicases. RNA helicases catalyze the unwinding of double-stranded RNA and play a role in RNA metabolism, including pre-mRNA splicing, ribosome biogenesis, and organellar gene expression.[supplied by OMIM
Sequence: SVGRTFCTMDGRGSPVHIHPSSALHEQETKLEWIIFHEVLVTTKVYARIVCPIRYEWVRDLLPKLHEFNAHDLSSVARREVREDARRRWTNKENVKQLKDGISKDVLKKMQRRNDDKSISDARARFSpecifications
| DHX40 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant DHX40. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q8IX18 | |
| DHX40 | |
| Recombinant protein corresponding to amino acids of human DHX40. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| DHX40 | |
| ARG147/DDX40/FLJ22060/FLJ34015/PAD | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 79665 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only