Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Dia 1, Polyclonal,

Catalog No. ABS12MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS12MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS12MI Supplier MilliporeSigma™ Supplier No. ABS12
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Dia 1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Protein diaphanous homolog 1, also known as Diaphanous-related formin-1 or DRF1, and encoded by the gene name DIAPH1 or DIAP1, is a homolog of Drosophila diaphanous, and it has been linked to autosomal dominant, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. DIAP1 has therefore been speculated to have a role in the regulation of actin polymerization in hair cells of the inner ear.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dia 1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O60610
Gene Symbols DIAPH1; DIAP1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the FH2 domain of human Dia 1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_005210
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.