Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Disc1 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Disc1.
Supplier: Abnova Corporation PAB12144
Specifications
Disc1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Disc1. | |
In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
Q811T9 | |
Rabbit | |
100 μL | |
Primary | |
Mouse, Rat | |
Liquid |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
Disc1 | |
Disc1 | |
A synthetic peptide corresponding to amino acids 700-800 of mouse Disc1. | |
RUO | |
244667 | |
Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction