Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dkk3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Dkk3.
Supplier: Abnova Corporation PAB8678
Specifications
Dkk3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Dkk3. | |
In PBS, pH 7.2 (30% glycerol, 0.01% thimerosal) | |
Reic/p29 | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects ∼38 KDa of Dkk3. | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
Dkk3 | |
Dkk3 | |
A synthetic peptide corresponding to residues surrounding amino acids 240 of rat Dkk3. | |
RUO | |
171548 | |
Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction